4,5-Dibromo-2-furoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,5-Dibromofuran-2-carboxylic acid

M-Clarity™ quality level = MQ100

656291-25G 656291-5G
SIAL656291-25GEA 306 GBP
SIAL656291-25G SIAL656291-5G
4,5-Dibromo-2-furoic acid, Sigma-Aldrich®
4,5-Dibromo-2-furoic acid
Formula: C₅H₂Br₂O₃
MW: 269.88 g/mol
MDL Number: MFCD00092311
CAS Number: 2434-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR