Benzoyl peroxide stabilised, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Stabilisation: Moistened with water
M-Clarity™ quality level = MQ200

517909-500G 517909-5G
SIAL517909-500GEA 198 GBP
SIAL517909-500G SIAL517909-5G
Benzoyl peroxide stabilised, Sigma-Aldrich®
Benzoyl peroxide
Formula: C₁₄H₁₀O₄
MDL Number: MFCD00003071
CAS Number: 94-36-0
EINECS: 202-327-6
UN: 3104
ADR: 5.2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR