Calcium trifluoromethanesulphonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Calcium trifluoromethanesulphonate, Calcium triflate

M-Clarity™ quality level = MQ200

515892-10G 515892-50G
SIAL515892-10GEA 188 GBP
SIAL515892-10G SIAL515892-50G
Calcium trifluoromethanesulphonate, Sigma-Aldrich®
Calcium trifluoromethanesulphonate
Formula: C₂CaF₆O₆S₂
MDL Number: MFCD00143911
CAS Number: 55120-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR