Potassium trifluoromethanesulphonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

422843-25G 422843-5G
SIAL422843-25GEA 145 GBP
SIAL422843-25G SIAL422843-5G
Potassium trifluoromethanesulphonate, Sigma-Aldrich®
Potassium trifluoromethanesulphonate
Formula: CF₃KO₃S
MW: 188.17 g/mol
MDL Number: MFCD00042370
CAS Number: 2926-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR