Dibutyltin diacetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Di-n-butyltin diacetate

M-Clarity™ quality level = MQ200

290890-250ML 290890-50ML
SIAL290890-250MLEA 258 GBP
SIAL290890-250ML SIAL290890-50ML
Dibutyltin diacetate, Sigma-Aldrich®
Dibutyltin diacetate
Formula: C₁₂H₂₄O₄Sn
MW: 351.03 g/mol
MDL Number: MFCD00008697
CAS Number: 1067-33-0
EINECS: 213-928-8
UN: 2788
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR