Diethylaluminium chloride 1 mol/l in hexane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: DEAC, Diethylaluminum chloride

M-Clarity™ quality level = MQ200

212806-100ML 212806-800ML
SIAL212806-100MLEA 127 GBP
SIAL212806-100ML SIAL212806-800ML
Diethylaluminium chloride 1 mol/l in hexane, Sigma-Aldrich®
Diethylaluminium chloride
Formula: C₄H₁₀AlCl
MW: 120.56 g/mol
Boiling Pt: 208
Melting Pt: -50
Density: 0.961
MDL Number: MFCD00000459
CAS Number: 96-10-6
EINECS: 202-477-2
UN: 3394
ADR: 4.2 I

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR