1-Acetylpiperazine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
023244-100G
023244-25G
FLUO023244-100GEA
0
GBP
FLUO023244-100G
FLUO023244-25G
1-Acetylpiperazine
1-Acetylpiperazine
Formula:
C₆H₁₂N₂O |
MDL Number:
MFCD00058676 CAS Number: 13889-98-0 EINECS: 237-659-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...