1-(3-Cyanobenzyl)piperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-[(Piperazin-1-yl)methyl]benzonitrile

021563-5G 021563-1G
FLUO021563-5GEA 0 GBP
FLUO021563-5G FLUO021563-1G
1-(3-Cyanobenzyl)piperazine
1-(3-Cyanobenzyl)piperazine
Formula: C₁₂H₁₅N₃
MW: 201.27 g/mol
CAS Number: 203047-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR