1-(4-Chlorobenzyl)piperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019041-5G 019041-25G
FLUO019041-5GEA 0 GBP
FLUO019041-5G FLUO019041-25G
1-(4-Chlorobenzyl)piperazine
1-(4-Chlorobenzyl)piperazine
Formula: C₁₁H₁₅ClN₂
MDL Number: MFCD00040791
CAS Number: 23145-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR