o-Toluic hydrazide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
o-Toluic acid hydrazide, 2-Methylbenzohydrazide
019002-10G
019002-5G
FLUO019002-10GEA
0
GBP
FLUO019002-10G
FLUO019002-5G
o-Toluic hydrazide
o-Toluic hydrazide
Formula:
C₈H₁₀N₂O MW: 150.18 g/mol |
MDL Number:
MFCD00014756 CAS Number: 7658-80-2 EINECS: 231-623-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...