1-(2-(Trifluoromethyl)phenyl)piperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018139-1G 018139-5G
FLUO018139-1GEA 0 GBP
FLUO018139-1G FLUO018139-5G
1-(2-(Trifluoromethyl)phenyl)piperazine
1-(2-(Trifluoromethyl)phenyl)piperazine
Formula: C₁₁H₁₃F₃N₂
MDL Number: MFCD02094727
CAS Number: 63854-31-9
EINECS: 264-514-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR