Zinc trifluoromethanesulphonate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Zinc(II) trifluoromethanesulphonate, Zinc triflate, Zinc trifluoromethanesulphonate
013959-25G
013959-5G
FLUO013959-25GEA
0
GBP
FLUO013959-25G
FLUO013959-5G
Zinc trifluoromethanesulphonate
Zinc trifluoromethanesulphonate
Formula:
C₂F₆O₆S₂Zn MW: 363.53 g/mol |
MDL Number:
MFCD00013229 CAS Number: 54010-75-2 EINECS: 258-922-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...