5-Fluoro-2-nitrobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013851-1G 013851-5G
FLUO013851-1GEA 0 GBP
FLUO013851-1G FLUO013851-5G
5-Fluoro-2-nitrobenzonitrile
5-Fluoro-2-nitrobenzonitrile
Formula: C₇H₃FN₂O₂
MDL Number: MFCD01632196
CAS Number: 50594-78-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR