3-Chlorophthalic anhydride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Chloroisobenzofuran-1,3-dione

013484-5G 013484-25G
FLUO013484-5GEA 0 GBP
FLUO013484-5G FLUO013484-25G
3-Chlorophthalic anhydride
3-Chlorophthalic anhydride
Formula: C₈H₃ClO₃
MW: 182.56 g/mol
MDL Number: MFCD00023107
CAS Number: 117-21-5
EINECS: 204-179-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR