2-Acetamido-5-nitropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013106-5G 013106-1G
FLUO013106-5GEA 0 GBP
FLUO013106-5G FLUO013106-1G
2-Acetamido-5-nitropyridine
2-Acetamido-5-nitropyridine
Formula: C₇H₇N₃O₃
MDL Number: MFCD00833973
CAS Number: 5093-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR