Scandium(III) trifluoromethanesulphonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Scandium(III) trifluoromethanesulphonate, Scandium triflate

009343-1G 009343-5G
FLUO009343-1GEA 0 GBP
FLUO009343-1G FLUO009343-5G
Scandium(III) trifluoromethanesulphonate
Scandium (III) trifluoromethanesulphonate
Formula: C₃F₉O₉S₃Sc
MDL Number: MFCD00192433
CAS Number: 144026-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR