1-Allylpiperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008909-1G 008909-5G
FLUO008909-1GEA 0 GBP
FLUO008909-1G FLUO008909-5G
1-Allylpiperazine
1-Allylpiperazine
Formula: C₇H₁₄N₂
MDL Number: MFCD00167970
CAS Number: 13961-36-9
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR