Ethyl 3,5-difluorobenzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 3,5-difluorobenzoate

008812-10G 008812-50G
FLUO008812-10GEA 0 GBP
FLUO008812-10G FLUO008812-50G
Ethyl 3,5-difluorobenzoate
Ethyl 3,5-difluorobenzoate
Formula: C₉H₈F₂O₂
MDL Number: MFCD03093801
CAS Number: 350-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR