Methyl-3,5-difluorobenzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3,5-difluorobenzoate

008811-1G 008811-5G 008811-25G
FLUO008811-1GEA 0 GBP
FLUO008811-1G FLUO008811-5G FLUO008811-25G
Methyl-3,5-difluorobenzoate
Methyl 3,5-difluorobenzoate
Formula: C₈H₆F₂O₂
MDL Number: MFCD01318160
CAS Number: 216393-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR