Methyl-3,4-difluorobenzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3,4-difluorobenzoate

008379-1G 008379-25G
FLUO008379-1GEA 0 GBP
FLUO008379-1G FLUO008379-25G
Methyl-3,4-difluorobenzoate
Methyl 3,4-difluorobenzoate
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD00276597
CAS Number: 369-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR