Potassium trifluoromethanesulphonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007227-25G 007227-100G
FLUO007227-25GEA 0 GBP
FLUO007227-25G FLUO007227-100G
Potassium trifluoromethanesulphonate
Potassium trifluoromethanesulphonate
Formula: CF₃KO₃S
MW: 188.17 g/mol
MDL Number: MFCD00042370
CAS Number: 2926-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR