Methyl-4-(trifluoromethyl)benzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006842-25G 006842-100G 006842-5G
FLUO006842-25GEA 0 GBP
FLUO006842-25G FLUO006842-100G FLUO006842-5G
Methyl-4-(trifluoromethyl)benzoate
Methyl 4-(trifluoromethyl)benzoate
Formula: C₉H₇F₃O₂
MDL Number: MFCD00042324
CAS Number: 2967-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR