Methyl-3-(trifluoromethyl)benzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006841-5G 006841-25G 006841-100G
FLUO006841-5GEA 0 GBP
FLUO006841-5G FLUO006841-25G FLUO006841-100G
Methyl-3-(trifluoromethyl)benzoate
Methyl 3-(trifluoromethyl)benzoate
Formula: C₉H₇F₃O₂
MDL Number: MFCD00043543
CAS Number: 2557-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR