Methyl-2-(trifluoromethyl)benzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-(trifluoromethyl)benzoate

006840-5G 006840-25G
FLUO006840-5GEA 0 GBP
FLUO006840-5G FLUO006840-25G
Methyl-2-(trifluoromethyl)benzoate
Methyl 2-(trifluoromethyl)benzoate
Formula: C₉H₇F₃O₂
MDL Number: MFCD00236050
CAS Number: 344-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR