Methyl-2-(trifluoromethoxy)benzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006838-1G 006838-5G
FLUO006838-1GEA 0 GBP
FLUO006838-1G FLUO006838-5G
Methyl-2-(trifluoromethoxy)benzoate
Methyl 2-(trifluoromethoxy)benzoate
Formula: C₉H₇F₃O₃
MDL Number: MFCD00052336
CAS Number: 148437-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR