Methyl-2,6-difluorobenzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2,6-difluorobenzoate

006792-5G 006792-25G 006792-100G
FLUO006792-5GEA 0 GBP
FLUO006792-5G FLUO006792-25G FLUO006792-100G
Methyl-2,6-difluorobenzoate
Methyl 2,6-difluorobenzoate
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD00051778
CAS Number: 13671-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR