Magnesium trifluoromethanesulphonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Magnesium trifluoromethanesulphonate, Magnesium triflate

006720-5G 006720-10G 006720-25G
FLUO006720-5GEA 0 GBP
FLUO006720-5G FLUO006720-10G FLUO006720-25G
Magnesium trifluoromethanesulphonate
Magnesium trifluoromethanesulphonate
Formula: C₂F₆MgO₆S₂
MDL Number: MFCD00042601
CAS Number: 60871-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR