Ethyl-3-(trifluoromethyl)benzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006146-5G 006146-25G 006146.100G
FLUO006146-5GEA 0 GBP
FLUO006146-5G FLUO006146-25G FLUO006146.100G
Ethyl-3-(trifluoromethyl)benzoate
Ethyl 3-(trifluoromethyl)benzoate
Formula: C₁₀H₉F₃O₂
MDL Number: MFCD00013560
CAS Number: 76783-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR