2-Fluoro-6-methoxypyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7907-1G PY-7907-5G
COBBPY-7907-1GEA 0 GBP
COBBPY-7907-1G COBBPY-7907-5G
2-Fluoro-6-methoxypyridine
2-Fluoro-6-methoxypyridine
Formula: C₆H₆FNO
MDL Number: MFCD09998161
CAS Number: 116241-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR