5-Fluoro-2-methoxypyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7847-1G PY-7847-5G PY-7847-25G
COBBPY-7847-1GEA 0 GBP
COBBPY-7847-1G COBBPY-7847-5G COBBPY-7847-25G
5-Fluoro-2-methoxypyridine
5-Fluoro-2-methoxypyridine
Formula: C₆H₆FNO
MDL Number: MFCD03095252
CAS Number: 51173-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR