3-Bromo-5-fluoro-2-methoxypyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7138-5G
PY-7138-1G
COBBPY-7138-5GEA
0
GBP
COBBPY-7138-5G
COBBPY-7138-1G
3-Bromo-5-fluoro-2-methoxypyridine
3-Bromo-5-fluoro-2-methoxypyridine
Formula:
C₆H₅BrFNO |
MDL Number:
MFCD04972397 CAS Number: 884494-81-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...