2-Acetamido-5-bromo-3-nitropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1266-5G PY-1266-1G
COBBPY-1266-5GEA 0 GBP
COBBPY-1266-5G COBBPY-1266-1G
2-Acetamido-5-bromo-3-nitropyridine
2-Acetamido-5-bromo-3-nitropyridine
Formula: C₇H₆BrN₃O₃
MDL Number: MFCD00770745
CAS Number: 381679-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR