3-Bromo-2-propoxypyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1236-5G PY-1236-1G
COBBPY-1236-5GEA 0 GBP
COBBPY-1236-5G COBBPY-1236-1G
3-Bromo-2-propoxypyridine
3-Bromo-2-propoxypyridine
Formula: C₈H₁₀BrNO
MDL Number: MFCD09743637
CAS Number: 760207-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR