3-Picolyl chloride hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Chloromethyl)-pyridinium chloride, 3-(Chloromethyl)pyridine hydrochloride

OS-7432-5G OS-7432-25G OS-7432-100G
COBBOS-7432-5GEA 0 GBP
COBBOS-7432-5G COBBOS-7432-25G COBBOS-7432-100G
3-Picolyl chloride hydrochloride
3-Picolyl chloride hydrochloride
Formula: C₆H₇Cl₂N
MDL Number: MFCD00012818
CAS Number: 6959-48-4
EINECS: 230-150-4
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR