2-Acetyl-7-bromobenzofuran
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1374-5G
HI-1374-1G
COBBHI-1374-5GEA
0
GBP
COBBHI-1374-5G
COBBHI-1374-1G
2-Acetyl-7-bromobenzofuran
2-Acetyl-7-bromobenzofuran
Formula:
C₁₀H₇BrO₂ |
MDL Number:
MFCD10699679 CAS Number: 460086-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...