5-Bromo-2-nitrobenzonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6919-5G
HC-6919-1G
HC-6919-10G
COBBHC-6919-5GEA
0
GBP
COBBHC-6919-5G
COBBHC-6919-1G
COBBHC-6919-10G
5-Bromo-2-nitrobenzonitrile
5-Bromo-2-nitrobenzonitrile
Formula:
C₇H₃BrN₂O₂ |
MDL Number:
MFCD10699674 CAS Number: 89642-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...