4-Bromo-3-nitrobenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6655-5G HC-6655-25G HC-6655-1G HC-6655-100G
COBBHC-6655-5GEA 0 GBP
COBBHC-6655-5G COBBHC-6655-25G COBBHC-6655-1G COBBHC-6655-100G
4-Bromo-3-nitrobenzonitrile
4-Bromo-3-nitrobenzonitrile
Formula: C₇H₃BrN₂O₂
MW: 227.02 g/mol
MDL Number: MFCD00982006
CAS Number: 89642-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR