2-Bromo-6-methoxypyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6404-5G
HC-6404-25G
HC-6404-100G
COBBHC-6404-5GEA
0
GBP
COBBHC-6404-5G
COBBHC-6404-25G
COBBHC-6404-100G
2-Bromo-6-methoxypyridine
2-Bromo-6-methoxypyridine
Formula:
C₆H₆BrNO MW: 188.02 g/mol |
MDL Number:
MFCD00088345 CAS Number: 40473-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...