5-Bromo-3-fluoro-2-methoxypyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2073-5G
HC-2073-1G
COBBHC-2073-5GEA
0
GBP
COBBHC-2073-5G
COBBHC-2073-1G
5-Bromo-3-fluoro-2-methoxypyridine
5-Bromo-3-fluoro-2-methoxypyridine
Formula:
C₆H₅BrFNO |
MDL Number:
MFCD04112553 CAS Number: 124432-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...