3-Bromo-5-isobutoxybenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4991-1G CA-4991-5G
COBBCA-4991-1GEA 0 GBP
COBBCA-4991-1G COBBCA-4991-5G
3-Bromo-5-isobutoxybenzoic acid
3-Bromo-5-isobutoxybenzoic acid
Formula: C₁₁H₁₃BrO₃
MDL Number: MFCD16618943
CAS Number: 1228956-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR