3-Bromo-5-hydroxybenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4961-5G CA-4961-25G CA-4961-1G
COBBCA-4961-5GEA 0 GBP
COBBCA-4961-5G COBBCA-4961-25G COBBCA-4961-1G
3-Bromo-5-hydroxybenzoic acid
3-Bromo-5-hydroxybenzoic acid
Formula: C₇H₅BrO₃
MDL Number: MFCD06797980
CAS Number: 140472-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR