Methyl-3-fluoro-4-hydroxybenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4328-5G CA-4328-1G
COBBCA-4328-5GEA 0 GBP
COBBCA-4328-5G COBBCA-4328-1G
Methyl-3-fluoro-4-hydroxybenzoate
Methyl 3-fluoro-4-hydroxybenzoate
Formula: C₈H₇FO₃
MDL Number: MFCD00215838
CAS Number: 403-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR