3-Chloro-5-nitrobenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4299-5G CA-4299-1G
COBBCA-4299-5GEA 0 GBP
COBBCA-4299-5G COBBCA-4299-1G
3-Chloro-5-nitrobenzonitrile
3-Chloro-5-nitrobenzonitrile
Formula: C₇H₃ClN₂O₂
MDL Number: MFCD00463805
CAS Number: 34662-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR