Methyl-4-bromo-2-fluorobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4291-1G CA-4291-25G CA-4291-5G CA-4291-100G
COBBCA-4291-1GEA 0 GBP
COBBCA-4291-1G COBBCA-4291-25G COBBCA-4291-5G COBBCA-4291-100G
Methyl-4-bromo-2-fluorobenzoate
Methyl 4-bromo-2-fluorobenzoate
Formula: C₈H₆BrFO₂
MDL Number: MFCD06203731
CAS Number: 179232-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR