Methyl-2,3-difluorobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2,3-difluorobenzoate

CA-4156-5G CA-4156-1G
COBBCA-4156-5GEA 0 GBP
COBBCA-4156-5G COBBCA-4156-1G
Methyl-2,3-difluorobenzoate
Methyl 2,3-difluorobenzoate
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD06203674
CAS Number: 18355-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR