5-Fluoro-2-nitrobenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1042-5G AN-1042-25G AN-1042-1G
COBBAN-1042-5GEA 0 GBP
COBBAN-1042-5G COBBAN-1042-25G COBBAN-1042-1G
5-Fluoro-2-nitrobenzonitrile
5-Fluoro-2-nitrobenzonitrile
Formula: C₇H₃FN₂O₂
MDL Number: MFCD01632196
CAS Number: 50594-78-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR