D-α-Aminoadipic acid
Supplier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
D-2-Aminoadipic acid, (R)-2-Aminohexanedioic acid, H-D-Aad-OH
4030344.0001
4030344.0005
BACE4030344.0001EA
0
GBP
BACE4030344.0001
BACE4030344.0005
D-α-Aminoadipic acid
D-α-Aminoadipic acid
Formula:
C₆H₁₁NO₄ |
MDL Number:
MFCD00063118 CAS Number: 7620-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...