L-α-Aminoadipic acid

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4014920.0001 4014920.0005 4014920.0025
BACE4014920.0001EA 0 GBP
BACE4014920.0001 BACE4014920.0005 BACE4014920.0025
L-α-Aminoadipic acid
L-α-Aminoadipic acid
Formula: C₆H₁₁NO₄
MDL Number: MFCD00002636
CAS Number: 1118-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR