Aluminium trifluoromethanesulphonate

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Aluminum trifluoromethanesulphonate, Aluminum triflate, Aluminum tris(trifluoromethanesulphonate)

38568.09 38568.18
38568.09EA 83.7 GBP
38568.09 38568.18
Aluminium trifluoromethanesulphonate
Aluminium trifluoromethanesulphonate

CAS No.: 74974-61-1 Molecular Formula: C3AlF9O9S3 Formula Weight: 474.20 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: ca 300° UN No.: 3261 MDL No.: MFCD00143596

Formula: C₃AlF₉O₉S₃
Melting Pt: ∼ 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00143596
CAS Number: 74974-61-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR