Cyclopropylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
003712-50G
003712-250G
FLUO003712-50GEA
0
GBP
FLUO003712-50G
FLUO003712-250G
Cyclopropylamine
Cyclopropylamine
Formula:
C₃H₇N Boiling Pt: 49 °C (1013 hPa) Melting Pt: –10 °C Density: 0,8131 g/cm³ (20 °C) Flash Pt: 1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001301 CAS Number: 765-30-0 EINECS: 212-142-2 UN: 2733 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...